missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1,2,3,4-tetrahydroisoquinoline, 95%

Product Code. 15451289
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451289 5 g 5g
15441289 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451289 Supplier Thermo Scientific Alfa Aesar Supplier No. H64191.06
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 4965-09-7 | C9H8NO6 | 226.17 g/mol

1-Methyl-1,2,3,4-tetrahydroisoquinoline (1MeTIQ) is an endogenous antidepressant and parkinsonism-preventing substance that demonstrates neuroprotectiveactivity. Following systemic administration in rats, 1-Methyl-1,2,3,4-tetrahydroisoquinoline produces antidepressant-like effect similar to the effect of imipramine. 1MeTIQ is a reversible short-acting moderate inhibitor of MAO A/B. Also known as 1,2,3,4-Tetrahydro-1-methylisoquinoline, 1MeTIQ. It is an important raw material and intermediate for revaprazan and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4965-09-7
MDL Number MFCD00798989

Specifications

Color Yellow
Boiling Point 119°C (18 mmHg)
Molecular Formula C9H8NO6
Quantity 5 g
Beilstein 82140
Synonym 4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid
Solubility Information Slightly soluble in water.
InChI Key WWCMFGBGMJAJRX-UHFFFAOYSA-M
SMILES COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC
Molecular Weight (g/mol) 226.17
PubChem CID 78690
Formula Weight 147.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Methyl-1,2,3,4-tetrahydroisoquinoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.