missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1,2-dihydropyrimidin-2-one, TRC

Product Code. 30464964
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464964 500 mg 500mg
30479059 100 mg 100mg
30478952 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464964 Supplier Toronto Research Chemicals Supplier No. TRCM336413500MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3739-81-9
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
Synonym 1-Methyl-2-pyrimidinone, 1-Methyl-2-pyrimidone, NSC 193514, 1-Methyl-2(1H)-pyrimidinone
IUPAC Name 1-methylpyrimidin-2-one
SMILES CN1C=CC=NC1=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H6N2O/c1-7-4-2-3-6-5(7)8/h2-4H,1H3
Formula Weight 110.048
Chemical Name or Material 1-Methyl-1,2-dihydropyrimidin-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.