missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-phenylhydrazine, 96%

Product Code. 10708742 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10708742 25 g Glass bottle 25g
10162911 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10708742 Supplier Thermo Scientific Acros Supplier No. 354790250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 618-40-6 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-40-6
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007627
InChI Key MWOODERJGVWYJE-UHFFFAOYSA-N
Synonym 1-methylphenylhydrazine, hydrazine, 1-methyl-1-phenyl, n-methyl-n-phenylhydrazine, n-phenyl-n-methylhydrazine, unii-yhb9czb63u, 1-methyl-1-phenyl-hydrazine, alpha-methyl-alpha-phenylhydrazine, hydrazine, methylphenyl, yhb9czb63u, .alpha.-methyl-.alpha.-phenylhydrazine
PubChem CID 36299
IUPAC Name 1-methyl-1-phenylhydrazine
SMILES CN(C1=CC=CC=C1)N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 1.0400g/mL
Boiling Point 75°C to 78°C (0.4 kPa)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C6H5N(CH3)NH2
Refractive Index 1.5810 to 1.585
Quantity 25 g
Beilstein 15, 117
Specific Gravity 1.04
Solubility Information Solubility in water: sparingly soluble in water.,miscible in ethanol,ether,benzene,chloroform
Formula Weight 122.17
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-phenylhydrazine, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-1-phenylhydrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.