missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-carboxylic acid, 97%

Product Code. 11956221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11956221 1 g 1g
11966221 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11956221 Supplier Thermo Scientific Acros Supplier No. 444080010
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5952-92-1 | C5H6N2O2 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5952-92-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.11
InChI Key UPPPWUOZCSMDTR-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-4-carboxylic acid, 1-methyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-methyl, 4-carboxy-1-methyl-1h-pyrazole, 1-methyl-pyrazole-4-carboxylic acid, 1-methylpyrazole-4-carboxylicacid, zlchem 1243, pubchem16306, pubchem20860, acmc-209me3
PubChem CID 643160
ChEBI CHEBI:74741
IUPAC Name 1-methylpyrazole-4-carboxylic acid
SMILES CN1C=C(C=N1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 207°C to 209°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 126.11
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 1-Methyl-1H-pyrazole-4-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-1H-pyrazole-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.