missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol, TRC

Product Code. 30478033
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478033 250 mg 250mg
30474167 25 mg 25mg
30483331 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478033 Supplier Toronto Research Chemicals Supplier No. TRCM337703250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122431-37-2
Molecular Formula C5H5N2OF3
Molecular Weight (g/mol) 166.1
Synonym Pyroxasulfone Impurity 11
IUPAC Name 2-methyl-5-(trifluoromethyl)pyrazol-3-ol
SMILES Cn1nc(cc1O)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H5F3N2O/c1-10-4(11)2-3(9-10)5(6,7)8/h2,11H,1H3
Formula Weight 166.0354
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.