missing translation for 'onlineSavingsMsg'
Learn More

1-Methyladenine, MedChemExpress

Product Code. 16437507
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437507 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16437507 Supplier Medchem Express Supplier No. HY113306
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 1-Methyladenine is a product of alkylation damage in DNA which can be repaired by damage reversal by oxidative demethylation.

1-Methyladenine is a product of alkylation damage in DNA which can be repaired by damage reversal by oxidative demethylation.

Specifications

Chemical Name or Material 1-Methyladenine
Physical Form Solid
CAS 5142-22-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C6H7N5
Solubility Information H2O : 5.2 mg/mL (34.86 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 5 mg/mL (33.52 mM; Need ultrasonic)
SMILES NC1=C2N=CN=C2N=CN1C
Molecular Weight (g/mol) 149.15
Formula Weight 149.15
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.