missing translation for 'onlineSavingsMsg'
Learn More

1-(Methylsulfonyl)-4-nitrobenzene, 95%, Thermo Scientific™

Product Code. 10161222
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10161222 1 g Amber Glass Bottle 1g
10405332 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10161222 Supplier Thermo Scientific Maybridge Supplier No. RH00770DA
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2976-30-9
Molecular Formula C7H7NO4S
Molecular Weight (g/mol) 201.196
MDL Number MFCD00047803
InChI Key XONGBDXIFQIQBN-UHFFFAOYSA-N
Synonym 1-methylsulfonyl-4-nitrobenzene, methyl 4-nitrophenyl sulfone, sulfone, methyl p-nitrophenyl, 1-methanesulfonyl-4-nitrobenzene, 4-methylsulfonylnitrobenzene, methyl p-nitrophenyl sulfone, 4-methylsulfonyl nitrobenzene, benzene, 1-methylsulfonyl-4-nitro, 4-methylsulfonyl nitro benzene, 1-methylsulphonyl-4-nitrobenzene
PubChem CID 258434
IUPAC Name 1-methylsulfonyl-4-nitrobenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula CH3SO2C6H4NO2
Quantity 1 g
Formula Weight 201.20
Percent Purity 97%
Chemical Name or Material 1-(Methylsulfonyl)-4-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.