missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-4-iodobenzene, 98%, Thermo Scientific Chemicals

Product Code. 11447057
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11447057

Brand: Thermo Scientific Alfa Aesar A14421.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as an active pharmaceutical intermediate. It is also used in the synthesis and characterization of photoelectronic polymers containing triphenylamine moiety and in the palladium-catalyzed conversion of aryl and vinyl triflates to bromides and chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate. It is also used in the synthesis and characterization of photoelectronic polymers containing triphenylamine moiety and in the palladium-catalyzed conversion of aryl and vinyl triflates to bromides and chlorides.

Solubility
Sparingly soluble in water at 25°C 3.6E-3 g/L.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20651-67-6
Molecular Formula C10H13I
Molecular Weight (g/mol) 260.118
MDL Number MFCD00051927
InChI Key XILRUONFYBUYIE-UHFFFAOYSA-N
Synonym 1-4'-iodophenyl butane, 4-n-butyliodobenzene, benzene, 1-butyl-4-iodo, 1-n-butyl-4-iodobenzene, 1-iodo-4-butylbenzene, 4-iodo-n-butylbenzene, 4-butyliodobenzene, p-iodo-n-butylbenzene, pubchem3969, 4-n-amyliodobenzene
PubChem CID 2724607
IUPAC Name 1-butyl-4-iodobenzene
SMILES CCCCC1=CC=C(C=C1)I

Specifications

Density 1.462
Boiling Point 80°C to 82°C (0.5 mmHg)
Refractive Index 1.569
Quantity 25 g
Beilstein 3030029
Sensitivity Light sensitive
Solubility Information Sparingly soluble in water at 25°C 3.6E-3g/L.
Formula Weight 260.12
Percent Purity 98%
Chemical Name or Material 1-n-Butyl-4-iodobenzene

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.