missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthalenemethylamine, 98+%

Product Code. 11372206
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11372206

Brand: Thermo Scientific Alfa Aesar L03541.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-31-0 | C11H12N | 158.22 g/mol

1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.

Solubility
Miscible with ethanol, ether and carbon disulfide.

Notes
Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-31-0
Molecular Formula C11H12N
Molecular Weight (g/mol) 158.22
MDL Number MFCD00004048
InChI Key NVSYANRBXPURRQ-UHFFFAOYSA-O
Synonym 1-naphthalenemethylamine, 1-naphthalenemethanamine, 1-naphthylmethylamine, 1-aminomethyl naphthalene, 1-naphthalenemethyl amine, c-naphthalen-1-yl-methylamine, 1-naphthylmethanamine, 1-aminomethylnaphthalene, naphthylmethylamine, 1-naphthylmethyl amine hydrochloride
PubChem CID 8355
IUPAC Name naphthalen-1-ylmethanamine
SMILES [NH3+]CC1=C2C=CC=CC2=CC=C1

Specifications

Density 1.075
Boiling Point 289°C to 292°C
Flash Point >110°C (230°F)
Refractive Index 1.643
Quantity 1 g
UN Number UN2735
Beilstein 2206459
Sensitivity Air sensitive; May form a precipitate
Solubility Information Miscible with ethanol,ether and carbon disulfide.
Formula Weight 157.22
Percent Purity ≥98%
Chemical Name or Material 1-Naphthalenemethylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.