missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoic acid, 98%

Product Code. 11444003
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444003 100 g 100g
11434003 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444003 Supplier Thermo Scientific Alfa Aesar Supplier No. A10273.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 86-55-5 | C11H8O2 | 172.183 g/mol

1-Naphthoic acid is used as an intermediate for the synthesis of pharmaceuticals, photochemicals, plant growth hormones, dyes and other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthoic acid is used as an intermediate for the synthesis of pharmaceuticals, photochemicals, plant growth hormones, dyes and other organic compounds.

Solubility
Freely soluble in hot alcohol and ether. Slightly soluble in hot water.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-55-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00004007
InChI Key LNETULKMXZVUST-UHFFFAOYSA-N
Synonym 1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
PubChem CID 6847
ChEBI CHEBI:36466
IUPAC Name naphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)O

Specifications

Melting Point 160°C to 162°C
Density 1.398
Boiling Point >300°C
Flash Point 195°C (383°F)
Quantity 100 g
Beilstein 1908896
Merck Index 14,6381
Solubility Information Freely soluble in hot alcohol and ether. Slightly soluble in hot water.
Formula Weight 172.18
Percent Purity 98%
Chemical Name or Material 1-Naphthoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.