missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoic acid, 98%

Product Code. p-170904 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10081470

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-55-5 | C11H8O2 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-55-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.18
MDL Number MFCD00004007
InChI Key LNETULKMXZVUST-UHFFFAOYSA-N
Synonym 1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
PubChem CID 6847
ChEBI CHEBI:36466
IUPAC Name naphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 158.0°C to 162.0°C
Color Brown to Yellow
Boiling Point 300.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C10H7CO2H
Quantity 25 g
Beilstein 09, 647
Merck Index 15, 6466
Solubility Information Solubility in water: slightly soluble in hot water. Other solubilities: freely soluble in hot alcohol and ether
Formula Weight 172.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Naphthoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Naphthoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.