missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol, Certified AR for Analysis, Fisher Chemical

Product Code. 10783991 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10783991 100 g Glass Jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10783991 Supplier Fisher Chemical Supplier No. N/0350/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 90-15-3
Melting Point 95°C
Color Beige
Boiling Point 278°C
Packaging Glass Jar
Molecular Formula C10H8O
MDL Number 3930
Quantity 100 g
Synonym 1-naphthol, 1-naphthalenol, alpha-naphthol, 1-hydroxynaphthalene, naphthol, fouramine ern, fourrine ern, tertral ern, furro er, basf ursol ern
InChI Key KJCVRFUGPWSIIH-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2O
IUPAC Name naphthalen-1-ol
Molecular Weight (g/mol) 144.173
PubChem CID 7005
ChEBI CHEBI:10319
Formula Weight 144.17g/mol
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Naphthol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.