missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthol, TRC

Product Code. 30138627
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138627 100 g 100g
30136751 10 g 10g
30040472 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138627 Supplier Toronto Research Chemicals Supplier No. TRCN367990100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-15-3
Formula Weight 144.0575
InChI Formula InChI=1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H
IUPAC Name naphthalen-1-ol
Molecular Formula C10 H8 O
Molecular Weight (g/mol) 144.17
Percent Purity >95%
SMILES Oc1cccc2ccccc12
Synonym Naphthalen-1-ol, 1-Naphthol, Duloxetine Hydrochloride Imp. D (Pharmeuropa), 1-Naphthalenol, 1-Naphthol (8CI), 1-Hydroxynaphthalene, 1-Naphthyl alcohol, BASF Ursol ERN, C.I. 76605, C.I. Oxidation Base 33, Durafur Developer D, Fouramine ERN, Fourrine 99, Fourrine ERN, Furro ER, NSC 9586, Nako TRB, Naphthol-1, Naphthyl-1-ol, Tertral ERN, Ursol ERN, Zoba ERN, α-Hydroxynaphthalene, α-Naphthol, α-Naphthyl alcohol
Chemical Name or Material 1-Naphthol
Grade HPLC
Show More Show Less

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.