missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl isothiocyanate, 98%

Product Code. 10437520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10437520 10 g Glass bottle 10g
10723711 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10437520 Supplier Thermo Scientific Acros Supplier No. 128240100
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 551-06-4 | C11H7NS | 185.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 551-06-4
Molecular Formula C11H7NS
Molecular Weight (g/mol) 185.25
MDL Number MFCD00003882
InChI Key JBDOSUUXMYMWQH-UHFFFAOYSA-N
Synonym 1-naphthyl isothiocyanate, 1-naphthylisothiocyanate, anit, kesscocide, alpha-naphthyl isothiocyanate, naphthalene, 1-isothiocyanato, naphthalene, isothiocyanato, isothiocyanic acid, 1-naphthyl ester, 1-naftylisothiokyanat, isothiocyanic acid 1-naphthyl ester
PubChem CID 11080
ChEBI CHEBI:35455
IUPAC Name 1-isothiocyanatonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55.5°C to 57.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C10H7NCS
Quantity 10 g
Beilstein 12, 1244
Fieser 01,718
Merck Index 15, 6493
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ether, benzene, hot alcohol,, acetone and ccl4
Formula Weight 185.25
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Naphthyl isothiocyanate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Naphthyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.