missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl phosphate, disodium salt hydrate, 98%

Product Code. 10482054 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10482054 1 g Glass bottle 1g
10011574 5 g Glass bottle 5g
10585924 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10482054 Supplier Thermo Scientific Acros Supplier No. 398260010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 207569-06-0 | C10H7Na2O4P | 268.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207569-06-0 , 2183-17-7
Molecular Formula C10H7Na2O4P
Molecular Weight (g/mol) 268.12
MDL Number MFCD00041007
InChI Key QYURIFWAOPAPAJ-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate disodium salt hydrate, sodium naphthalen-1-yl phosphate hydrate, disodium 1-naphthyl phosphate hydrate, disodium hydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h7o4p.2na.h2o, 1-naphthyl?phosphate?disodium?salt?hydrate, 1-naphthyl phosphate disodium salt monohydrate, dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt
PubChem CID 24212014
IUPAC Name disodium;naphthalen-1-yl phosphate;hydrate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 286.13
Percent Purity 98%
Chemical Name or Material 1-Naphthyl phosphate, disodium salt hydrate
compliance-icons
Product Identifier
  • 1-Naphthyl phosphate, disodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.