missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-2-(n-octyloxy)benzene, 98%

Product Code. 11483557
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483557 25 g 25g
11473557 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483557 Supplier Thermo Scientific Alfa Aesar Supplier No. A17320.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 37682-29-4 | C14H21NO3 | 251.326 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37682-29-4
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.326
MDL Number MFCD00014693
InChI Key CXVOIIMJZFREMM-UHFFFAOYSA-N
Synonym 2-nitrophenyl octyl ether, 1-nitro-2-octyloxy benzene, octyl o-nitrophenyl ether, 2-nitrophenyl n-octyl ether, 2-octyloxy nitrobenzene, benzene, 1-nitro-2-octyloxy, 1-2-nitrophenoxy octane, 1-nitro-2-octyloxybenzene, 2-n-octyloxy nitrobenzene, npoe
PubChem CID 169952
IUPAC Name 1-nitro-2-octoxybenzene
SMILES CCCCCCCCOC1=CC=CC=C1[N+](=O)[O-]

Specifications

Density 1.04
Boiling Point 163°C to 165°C (2 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.511
Quantity 25 g
Beilstein 2529905
Formula Weight 251.32
Percent Purity 98%
Chemical Name or Material 1-Nitro-2-(n-octyloxy)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.