missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-ol, 98%

Product Code. 11489982
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489982 10 g 10g
11499982 50 g 50g
11409992 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489982 Supplier Thermo Scientific Alfa Aesar Supplier No. A12079.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3391-86-4 | C8H16O | 128.215 g/mol

Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3391-86-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00004589
InChI Key VSMOENVRRABVKN-UHFFFAOYSA-N
Synonym 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
PubChem CID 18827
ChEBI CHEBI:34118
IUPAC Name oct-1-en-3-ol
SMILES CCCCCC(C=C)O

Specifications

Melting Point -49°C
Density 0.833
Boiling Point 173°C to 175°C
Flash Point 61°C (141°F)
Odor Characteristic
Refractive Index 1.437
Quantity 10 g
Beilstein 1744110
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 1-Octen-3-ol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octen-3-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Octen-3-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.