missing translation for 'onlineSavingsMsg'
Learn More

1-(p-Toluenesulfonyl)imidazole, 99%

Product Code. 10052970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10052970 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10052970 Supplier Thermo Scientific Acros Supplier No. 223300050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2232-08-8 | C10H10N2O2S | 222.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2232-08-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 75°C to 80°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C10H10N2O2S
MDL Number MFCD00005285
Quantity 5 g
Synonym 1-p-toluenesulfonyl imidazole, 1-tosyl-1h-imidazole, n-tosylimidazole, 1-4-methylphenyl sulfonyl-1h-imidazole, 1-tosylimidazole, 1-p-toluenesulphonyl imidazole, 1-p-tolylsulfonyl imidazole, 1h-imidazole, 1-4-methylphenyl sulfonyl, 1-toluene-p-sulphonyl imidazole, 1-4-methylbenzenesulfonyl imidazole
InChI Key YJYMYJRAQYREBT-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=CN=C1
IUPAC Name 1-(4-methylphenyl)sulfonylimidazole
Molecular Weight (g/mol) 222.26
PubChem CID 75219
Formula Weight 222.26
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-(p-Toluenesulfonyl)imidazole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-(p-Toluenesulfonyl)imidazole
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.