missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt monohydrate, 98+%, HPLC grade
GREENER_CHOICE

Product Code. 10178230 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10178230 25 g Glass Bottle 25g
10485661 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Artikelnummer. 10178230 Leverandør Thermo Scientific Leverandørnr. 423200250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
Denne vare kan ikke returneres. Se returpolicy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

CAS: 207605-40-1 | C5H11NaO3S·H2O | 192.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 207605-40-1
Molecular Formula C5H11NaO3S·H2O
Molecular Weight (g/mol) 192.22
MDL Number MFCD00149548
InChI Key FPQYXAFKHLSWTI-UHFFFAOYSA-M
Synonym sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade
PubChem CID 23693099
IUPAC Name sodium;pentane-1-sulfonate;hydrate
SMILES CCCCCS(=O)(=O)[O-].O.[Na+]

Tekniske data

Absorbance (0.25M aq. soln.) (1 cm cell vs HPLC water), 0.05 max. at 220nm, 0.03 max. at 230nm, 0.02 max. at 240nm, 0.01 max. at 250nm, 0.01 max. at 260nm
Melting Point >300°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula CH3(CH2)4SO3Na·H2O
Quantity 25 g
Beilstein 04, III, 23
Solubility Information Solubility in water: soluble
Formula Weight 192.22
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Pentanesulfonic Acid, Sodium Salt Monohydrate
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 1-Pentanesulfonic acid, sodium salt monohydrate
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.