missing translation for 'onlineSavingsMsg'
Learn More

1-Pentyne, 99%

Product Code. 11307725
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307725 5 g 5g
11317725 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307725 Supplier Thermo Scientific Alfa Aesar Supplier No. L02479.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-19-0 | C5H8 | 68.12 g/mol

1-Pentyne has been used in preparation of:lithium acetylides, required for asymmetric synthesis of α,α-dibranched propargyl sulfinamides. It is also used to synthesize 7-hydroxy-10-methoxy-3H-naphtho[2.1-b]pyrans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentyne has been used in preparation of:lithium acetylides, required for asymmetric synthesis of α,α-dibranched propargyl sulfinamides. It is also used to synthesize 7-hydroxy-10-methoxy-3H-naphtho[2.1-b]pyrans.

Solubility
Soluble in water. (1.05 g/L) at 20°C.

Notes
Store at 2-8°C. Incompatible with oxidizing agents. Store away from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-19-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.12
MDL Number MFCD00009469
InChI Key IBXNCJKFFQIKKY-UHFFFAOYSA-N
Synonym 1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3
PubChem CID 12309
IUPAC Name pent-1-yne
SMILES CCCC#C

Specifications

Melting Point -106°C
Density 0.692
Boiling Point 39°C to 41°C
Flash Point −20°C (−4°F)
Odor Unpleasant
Linear Formula CH3CH2CH2C≡CH
Refractive Index 1.386
Quantity 5 g
UN Number UN3295
Beilstein 1697133
Solubility Information Soluble in water. (1.05g/L) at 20°C.
Formula Weight 68.12
Percent Purity 99%
Chemical Name or Material 1-Pentyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.