missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1,2-propanedione-2-oxime, 99%

Product Code. 10726803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10726803 10 g Glass bottle 10g
10551841 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10726803 Supplier Thermo Scientific Acros Supplier No. 205020100
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 119-51-7 | C9H9NO2 | 163.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-51-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00002115
InChI Key YPINLRNGSGGJJT-JXMROGBWSA-N
Synonym 1-phenyl-1,2-propanedione-2-oxime, 2-hydroxyimino propiophenone, isonitrosopropiophenone, 2-hydroxyiminopropiophenone, 2-hydroxyimino-1-phenylpropan-1-one, propiophenone, isonitroso, 2-isonitrosopropiophenone, alpha-oximinopropiophenone, 1-phenyl-1,2-propanedione 2-oxime, .alpha.-oximinopropiophenone
PubChem CID 9566063
IUPAC Name (2E)-2-hydroxyimino-1-phenylpropan-1-one
SMILES C\C(=N/O)C(=O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 111.0°C to 115.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5COC(=NOH)CH3
Quantity 10 g
Beilstein 07, III, 3464
Solubility Information Solubility in water: 1.3g/L (20°C)
Formula Weight 163.18
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material 1-Phenyl-1, 2-propanedione-2-oxime, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.