missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1,2-propanedione, 98%

Product Code. 10472401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10472401 5 g Glass bottle 5g
10279493 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10472401 Supplier Thermo Scientific Acros Supplier No. 207360050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 579-07-7 | C9H8O2 | 148.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-07-7
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00008755
InChI Key BVQVLAIMHVDZEL-UHFFFAOYSA-N
Synonym 1-phenyl-1,2-propanedione, acetylbenzoyl, acetyl benzoyl, benzoylacetyl, methylphenylglyoxal, pyruvophenone, 1,2-propanedione, 1-phenyl, phenylmethyldiketone, benzoyl methyl ketone, methyl phenyl diketone
PubChem CID 11363
ChEBI CHEBI:63552
IUPAC Name 1-phenylpropane-1,2-dione
SMILES CC(=O)C(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.1000g/mL
Boiling Point 103.0°C to 105.0°C (14.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COCOCH3
Refractive Index 1.5300 to 1.5340
Quantity 5 g
Beilstein 07, 677
Specific Gravity 1.1
Solubility Information Solubility in water: insoluble
Formula Weight 148.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1, 2-propanedione, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenyl-1,2-propanedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.