missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenyl-1-propanol, 98+%

Product Code. 11449782
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449782 100 g 100g
11439782 25 g 25g
11459782 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449782 Supplier Thermo Scientific Alfa Aesar Supplier No. A10830.22
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-54-9 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-54-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004564
InChI Key DYUQAZSOFZSPHD-UHFFFAOYSA-N
Synonym 1-phenyl-1-propanol, phenycholon, fepar, ejibil, felicur, livonal, 1-phenylpropanol, gallenperlen, phenylchol, bilergon
PubChem CID 7147
IUPAC Name 1-phenylpropan-1-ol
SMILES CCC(C1=CC=CC=C1)O

Specifications

Density 0.994
Boiling Point 217°C to 219°C
Flash Point 95°C (203°F)
Refractive Index 1.52
Quantity 100 g
Beilstein 1906759
Merck Index 14,3768
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material (±)-1-Phenyl-1-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.