missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-propanol 99%, ACROS Organics™

Product Code. 10041640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
250g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10041640 250g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10041640 Supplier Thermo Scientific Acros Supplier No. 150032500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 93-54-9
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9900g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H12O
Linear Formula C2H5CH(C6H5)OH
Refractive Index 1.5190 to 1.5210 (20°C, 589nm)
MDL Number MFCD00004564
Quantity 250 g
Synonym 1-phenyl-1-propanol, phenycholon, fepar, ejibil, felicur, livonal, 1-phenylpropanol, gallenperlen, phenylchol, bilergon
InChI Key DYUQAZSOFZSPHD-UHFFFAOYSA-N
SMILES CCC(C1=CC=CC=C1)O
IUPAC Name 1-phenylpropan-1-ol
Molecular Weight (g/mol) 136.194
PubChem CID 7147
Formula Weight 136.19
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-propanol, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.