missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-propanol, 98%

Product Code. 10777973 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10777973 5 g Glass bottle 5g
10661784 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10777973 Supplier Thermo Scientific Acros Supplier No. 183060050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14898-87-4 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-87-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004540
InChI Key WYTRYIUQUDTGSX-UHFFFAOYNA-N
Synonym 1-phenyl-2-propanol, benzyl methyl carbinol, 2-hydroxy-1-phenylpropane, benzylmethylcarbinol, 2-propanol, 1-phenyl, benzeneethanol, .alpha.-methyl, alpha-methyl-phenethyl alcohol, benzeneethanol, alpha-methyl, alpha-methylbenzeneethanol, .alpha.-methylphenethyl alcohol
PubChem CID 94185
IUPAC Name 1-phenylpropan-2-ol
SMILES CC(O)CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9900g/mL
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH(OH)CH3
Refractive Index 1.5200 to 1.522
Quantity 5 g
Beilstein 06,503
Specific Gravity 0.99
Formula Weight 136.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-2-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenyl-2-propanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.