missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-propyn-1-one, 97%

Product Code. 15482162 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15482162 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15482162 Supplier Thermo Scientific Acros Supplier No. 456430010
 Restricted Item
Estimated Shipment: 11-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3623-15-2 | C9H6O | 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3623-15-2
Melting Point 51.0°C to 52.0°C
Boiling Point 90.0°C (0.8 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C9H6O
MDL Number MFCD06661646
Quantity 1 g
Synonym 1-phenyl-2-propyn-1-one, propiolophenone, benzoylacetylene, propynophenone, propioloylbenzene, ethyne, benzoyl, ethynyl phenyl ketone, phenyl ethynyl ketone, 3-oxo-3-phenylpropyne, 2-propyn-1-one, 1-phenyl
InChI Key JITPLZPWKYUTDM-UHFFFAOYSA-N
SMILES O=C(C#C)C1=CC=CC=C1
IUPAC Name 1-phenylprop-2-yn-1-one
Molecular Weight (g/mol) 130.15
PubChem CID 77179
Formula Weight 130.15
Percent Purity 97%
Chemical Name or Material 1-Phenyl-2-propyn-1-one
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenyl-2-propyn-1-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.