missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-pyrrolidinone, 99%

Product Code. 11373636
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373636 5 g 5g
11383636 25 g 25g
11393636 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373636 Supplier Thermo Scientific Alfa Aesar Supplier No. L14660.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4641-57-0 | C10H11NO | 161.20 g/mol

A novel derivative of 1-phenyl-2-pyrrolidinone, blebbistatin, inhibits non-muscle myocin II activity with high specificity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A novel derivative of 1-phenyl-2-pyrrolidinone, blebbistatin, inhibits non-muscle myocin II activity with high specificity.

Solubility
Soluble in chloroform and ethyl Acetate. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4641-57-0
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00003192
InChI Key JMVIVASFFKKFQK-UHFFFAOYSA-N
Synonym 1-phenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-phenyl, n-phenyl-2-pyrrolidone, n-phenyl-2-pyrrolidinone, 1-phenyl-pyrrolidin-2-one, 1-phenyl-2-pyrrolidone, n-phenylpyrrolidinone, n-phenyl pyrrolidone, pubchem23691, maybridge3_004960
PubChem CID 78375
IUPAC Name 1-phenylpyrrolidin-2-one
SMILES O=C1CCCN1C1=CC=CC=C1

Specifications

Melting Point 63°C to 68°C
Boiling Point 123°C (0.2 mmHg)
Odor Odorless
Quantity 5 g
Beilstein 130685
Solubility Information Soluble in chloroform and ethyl Acetate. Insoluble in water.
Formula Weight 161.2
Percent Purity 99%
Chemical Name or Material 1-Phenyl-2-pyrrolidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.