missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-thiourea, 97%

Product Code. 10036383 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10036383 100 g Glass bottle 100g
10616792 5 g Glass bottle 5g
10107703 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10036383 Supplier Thermo Scientific Acros Supplier No. 207251000
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-85-5 | C7H8N2S | 152.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-85-5
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.22
MDL Number MFCD00004933
InChI Key FULZLIGZKMKICU-UHFFFAOYSA-N
Synonym n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
PubChem CID 676454
ChEBI CHEBI:46261
IUPAC Name phenylthiourea
SMILES NC(=S)NC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 148°C to 150°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5NHCSNH2
Quantity 100 g
Beilstein 12, 388
Merck Index 15, 7424
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol,aq.ether,naoh,ethanol
Formula Weight 152.21
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-2-thiourea
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.