missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-trimethylsilylacetylene, 99%

Product Code. 15442248
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442248 5 g 5g
15452248 25 g 25g
15462248 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442248 Supplier Thermo Scientific Alfa Aesar Supplier No. H55866.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2170-06-1 | C11H14Si | 174.32 g/mol

1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

Solubility
Sparingly Soluble (6.1E-3 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2170-06-1
Molecular Formula C11H14Si
Molecular Weight (g/mol) 174.32
MDL Number MFCD00054860
InChI Key UZIXCCMXZQWTPB-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CC1=CC=CC=C1

Specifications

Color Colorless
Density 0.886 g/mL
Boiling Point 87°C to 88°C (9 mmHg)
Flash Point 74°C (165°F)
Linear Formula C6H5C≡CSi(CH3)3
Refractive Index 1.528
Quantity 5 g
Beilstein 3088678
Synonym 1-phenyl-2-trimethylsilyl acetylene, trimethyl phenylethynyl silane, silane, trimethyl phenylethynyl, 1-phenyl-2-trimethylsilylacetylene, phenylethynyltrimethylsilane, trimethylsilyl phenylacetylene, trimethyl 2-phenylethynyl silane, phenylethynyl-trimethylsilane, phenylethynyl trimethylsilane, acmc-1bika
Solubility Information Sparingly Soluble (6.1E-3 g/L) (25°C)
IUPAC Name trimethyl(2-phenylethynyl)silane
PubChem CID 137464
Formula Weight 174.32
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-2-trimethylsilylacetylene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenyl-2-trimethylsilylacetylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Phenyl-2-(trimethylsilyl)acetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.