missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-trimethylsilylacetylene, 99%

Product Code. 15452248
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15452248

Brand: Thermo Scientific Alfa Aesar H55866.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2170-06-1 | C11H14Si | 174.32 g/mol

1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized Pt-η-alkyne complexes.

Solubility
Sparingly Soluble (6.1E-3 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2170-06-1
Molecular Formula C11H14Si
Molecular Weight (g/mol) 174.32
MDL Number MFCD00054860
InChI Key UZIXCCMXZQWTPB-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CC1=CC=CC=C1

Specifications

Color Colorless
Density 0.886 g/mL
Boiling Point 87°C to 88°C (9 mmHg)
Flash Point 74°C (165°F)
Linear Formula C6H5C≡CSi(CH3)3
Refractive Index 1.528
Quantity 25 g
Beilstein 3088678
Synonym tetrakis tert-butoxy hafnium, hafnium; 2-methylpropan-2-ol
Solubility Information Sparingly Soluble (6.1E-3 g/L) (25°C)
IUPAC Name hafnium;2-methylpropan-2-ol
PubChem CID 25199745
Formula Weight 174.32
Percent Purity 99%
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.