missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-trimethylsilyl-2-propyn-1-ol, 97%

Product Code. 15430228
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430228 5 g 5g
15440228 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430228 Supplier Thermo Scientific Alfa Aesar Supplier No. H53426.06
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 89530-34-7 | C12H16OSi | 204.344 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89530-34-7
Molecular Formula C12H16OSi
Molecular Weight (g/mol) 204.344
MDL Number MFCD05864336
InChI Key RVTDALNDYLDVMN-UHFFFAOYSA-N
Synonym 1-phenyl-3-trimethylsilyl-2-propyn-1-ol, 1-phenyl-3-trimethylsilyl prop-2-yn-1-ol, benzenemethanol, a-2-trimethylsilyl ethynyl, acmc-20ln8h, alpha-trimethylsilyl ethynylbenzyl alcohol, 4,4-dimethyl-1-phenyl-4-silapent-2-yn-1-ol, alpha-trimethylsilyl ethynyl-benzenemethanol, alpha-trimethylsilyl-ethynyl-benzenemethanol, alpha-trimethylsilyl ethynyl-benzene-methanol, 1-phenyl-3-1,1,1-trimethylsilyl-2-propyn-1-ol
PubChem CID 2760416
IUPAC Name 1-phenyl-3-trimethylsilylprop-2-yn-1-ol
SMILES C[Si](C)(C)C#CC(C1=CC=CC=C1)O

Specifications

Quantity 5 g
Flash Point 110°C (230°F)
Formula Weight 204.34
Percent Purity 97%
Refractive Index 1.5193
Density 0.961
Chemical Name or Material 1-Phenyl-3-trimethylsilyl-2-propyn-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.