missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)-1H-pyrrole, 97%, Thermo Scientific™

Product Code. 10708625
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10708625 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10708625 Supplier Thermo Scientific Maybridge Supplier No. BTB07001DA
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16851-82-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H9NO2S
MDL Number MFCD00067739
Quantity 1 g
Synonym 1-phenylsulfonyl pyrrole, 1-phenylsulfonyl-1h-pyrrole, n-benzenesulphonylpyrrole, 1-benzenesulfonyl pyrrole, n-benzenesulfonylpyrrole, 1-benzenesulfonyl-1h-pyrrole, n-phenylsulfonylpyrrole, phenylsulfonyl pyrrole, 1h-pyrrole,1-phenylsulfonyl, 1h-pyrrole, 1-phenylsulfonyl
InChI Key PPPXRIUHKCOOMU-UHFFFAOYSA-N
SMILES O=S(=O)(N1C=CC=C1)C1=CC=CC=C1
IUPAC Name 1-(benzenesulfonyl)pyrrole
Molecular Weight (g/mol) 207.25
PubChem CID 140146
Formula Weight 207.25
Percent Purity 97%
Chemical Name or Material 1-(Phenylsulfonyl)-1H-pyrrole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.