missing translation for 'onlineSavingsMsg'
Learn More

1-Propionylpyrene, 98%, Thermo Scientific™

Product Code. 15107442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15107442 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15107442 Supplier Thermo Scientific Acros Supplier No. 449320010
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 500004-32-0
Melting Point 80.0°C to 82.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C19H14O
Quantity 1 g
Solubility Information Solubility in water: insoluble.
InChI Key COIWAOXGROSMQJ-UHFFFAOYSA-N
SMILES CCC(=O)C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1
IUPAC Name 1-pyren-1-ylpropan-1-one
Molecular Weight (g/mol) 258.31
PubChem CID 232861
Formula Weight 258.31
Percent Purity 98%
Chemical Name or Material 1-Propionylpyrene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.