missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl-3-ethylbenzene, 98%

Product Code. 11479846
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479846 25 g 25g
11469846 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479846 Supplier Thermo Scientific Alfa Aesar Supplier No. B20325.14
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14411-56-4 | C12H18 | 162.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14411-56-4
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00060845
InChI Key MUJPTTGNHRHIPH-UHFFFAOYSA-N
PubChem CID 139753
IUPAC Name 1-tert-butyl-3-ethylbenzene
SMILES CCC1=CC(=CC=C1)C(C)(C)C

Specifications

Density 0.86
Boiling Point 205°C
Odor Mild
Quantity 25 g
Synonym 1-t-butyl-3-ethylbenzene, 1-tert-butyl-3-ethylbenzene, benzene, 1-1,1-dimethylethyl-3-ethyl, benzene,1-1,1-dimethylethyl-3-ethyl, butylethylbenzene, m-tert-butyl-ethylbenzene, acmc-1cb13, 1-tert-butyl-3-ethyl-benzene, 1-1,1-dimethylethyl-3-ethylbenzene, 1-tert-butyl-3-ethylbenzene 5g
Formula Weight 162.27
Percent Purity 98%
Chemical Name or Material 1-tert-Butyl-3-ethylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.