missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl 4-(9-H-fluoren-9-ylmethyl) hydrogen (2R)-piperazine-1,2,4-tricarboxyl, 97%, Thermo Scientific™

Product Code. 10225984
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225984 1 g Amber Glass Bottle 1g
10759205 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225984 Supplier Thermo Scientific Maybridge Supplier No. MO08425DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 209593-18-0
Molecular Formula C25H28N2O6
Molecular Weight (g/mol) 452.507
MDL Number MFCD02179102
InChI Key ZVHNNCSUTNWKFC-OAQYLSRUSA-N
Synonym r-1-n-boc-4-n-fmoc-2-piperazine carboxylic acid, r-1-n-boc-4-n-fmoc-2-piperazinecarboxylicacid, r-4-9h-fluoren-9-yl methoxy carbonyl-1-tert-butoxycarbonyl piperazine-2-carboxylic acid, 1-boc-4-fmoc-piperazine-2-r-carboxylic acid, 2r-1-tert-butoxycarbonyl-4-9h-fluoren-9-ylmethoxy carbonyl piperazine-2-carboxylic acid, r-piperazine-1,2,4-tricarboxylic acid 1-tert-butyl ester 4-9h-fluoren-9-ylmethyl ester, 1-tert-butyl 4-9-h-fluoren-9-ylmethyl hydrogen 2r-piperazine-1,2,4-tricarboxylate, bestipharma 115-414, r-4-fmoc-1-tert-butoxycarbonyl piperazine-2-carboxylic acid, 2r-4-9h-fluoren-9-ylmethoxycarbonyl-1-2-methylpropan-2-yl oxycarbonyl piperazine-2-carboxylic acid
PubChem CID 7128295
IUPAC Name (2R)-4-(9H-fluoren-9-ylmethoxycarbonyl)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperazine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCN(CC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 452.5
Percent Purity 97%
Chemical Name or Material 1-tert-Butyl 4-(9-H-fluoren-9-ylmethyl) hydrogen (2R)-piperazine-1, 2, 4-tricarboxyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.