missing translation for 'onlineSavingsMsg'
Learn More

(1-Tetradecyl)trimethylammonium bromide, 98%

Product Code. 11338355
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338355 500 g 500g
11318355 25 g 25g
11328355 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338355 Supplier Thermo Scientific Alfa Aesar Supplier No. L10294.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1119-97-7 | C17H38BrN | 336.40 g/mol

Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.

Solubility
Soluble in water. (100 g/L)

Notes
Light sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-97-7
Molecular Formula C17H38BrN
Molecular Weight (g/mol) 336.40
MDL Number MFCD00011770
InChI Key CXRFDZFCGOPDTD-UHFFFAOYSA-M
Synonym cetrimide, tetradecyltrimethylammonium bromide, myristyltrimethylammonium bromide, tetradonium bromide, n,n,n-trimethyltetradecan-1-aminium bromide, tetradecyl trimethyl ammonium bromide, myrtrimonium bromide, mytab, mitmab, morpan t
PubChem CID 14250
ChEBI CHEBI:3565
IUPAC Name trimethyl(tetradecyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point 245°C to 250°C
Odor Odorless
Linear Formula CH3(CH2)13N(CH3)3Br
Quantity 500 g
UN Number UN1759
Beilstein 3633227
Merck Index 14,6336
Solubility Information Soluble in water. (100g/L)
Formula Weight 336.4
Percent Purity 98%
Chemical Name or Material (1-Tetradecyl)trimethylammonium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.