missing translation for 'onlineSavingsMsg'
Learn More

1-Thionaphthol, 99%

Product Code. 11461327
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11461327

Brand: Thermo Scientific Alfa Aesar B24876.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 529-36-2 | C10H7S | 159.23 g/mol

1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.

Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-36-2
Molecular Formula C10H7S
Molecular Weight (g/mol) 159.23
MDL Number MFCD00039599
InChI Key SEXOVMIIVBKGGM-UHFFFAOYSA-M
Synonym 1-naphthalenethiol, 1-thionaphthol, naphthalenethiol, 1-naphthylthiol, unii-9a78cp495h, 1-mercaptonaphthalene, .alpha.-naphthyl mercaptan, naphthalene thiol, 1-napthalenethiol, naphthalen-1-thiol
PubChem CID 68259
IUPAC Name naphthalene-1-thiol
SMILES [S-]C1=C2C=CC=CC2=CC=C1

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.
Formula Weight 160.24
Percent Purity 99%
Chemical Name or Material 1-Thionaphthol
compliance-icons
Product Identifier
  • 1-Thionaphthol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.