missing translation for 'onlineSavingsMsg'
Learn More

1-Thionaphthol, 99%

Product Code. 11481327
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481327 25 g 25g
11461327 1 g 1g
11471327 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11481327 Fornitore Thermo Scientific Alfa Aesar N. del fornitore B24876.14
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 529-36-2 | C10H7S | 159.23 g/mol

1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.

Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 529-36-2
Molecular Formula C10H7S
Molecular Weight (g/mol) 159.23
MDL Number MFCD00039599
InChI Key SEXOVMIIVBKGGM-UHFFFAOYSA-M
Synonym 1-naphthalenethiol, 1-thionaphthol, naphthalenethiol, 1-naphthylthiol, unii-9a78cp495h, 1-mercaptonaphthalene, .alpha.-naphthyl mercaptan, naphthalene thiol, 1-napthalenethiol, naphthalen-1-thiol
PubChem CID 68259
IUPAC Name naphthalene-1-thiol
SMILES [S-]C1=C2C=CC=CC2=CC=C1

Specifica

Quantity 25 g
Sensitivity Air Sensitive
Solubility Information Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.
Formula Weight 160.24
Percent Purity 99%
Chemical Name or Material 1-Thionaphthol
compliance-icons
Identificativo prodotto
  • 1-Thionaphthol
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.