missing translation for 'onlineSavingsMsg'
Learn More

1-Tritylimidazole-4-carboxaldehyde, 98%

Product Code. 11470838
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470838 25 g 25g
11450838 1 g 1g
11460838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470838 Supplier Thermo Scientific Alfa Aesar Supplier No. B25272.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33016-47-6 | C23H18N2O | 338.41 g/mol

1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33016-47-6
Molecular Formula C23H18N2O
Molecular Weight (g/mol) 338.41
MDL Number MFCD02179554
InChI Key YQYLLBSWWRWWAY-UHFFFAOYSA-N
Synonym 1-tritylimidazole-4-carboxaldehyde, 1-trityl-1h-imidazole-4-carbaldehyde, 1-trityl-1h-imidazole-4-carboxaldehyde, 1-trityl-imdazole-4-carboxadehyde, 4-formyl-1-tritylimidazole, 4-formyl-1-trityl-1h-imidazole, 1-trityl-4-formylimidazole, 1-triphenylmethyl imidazole-4-carbaldehyde, 1-triphenylmethyl-1h-imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 1-triphenylmethyl
PubChem CID 618233
SMILES O=CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼192°C (decomposition)
Quantity 25 g
Sensitivity Air Sensitive
IUPAC Name 1-(triphenylmethyl)-1H-imidazole-4-carbaldehyde
Formula Weight 338.41
Percent Purity 98%
Chemical Name or Material 1-Tritylimidazole-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.