missing translation for 'onlineSavingsMsg'
Learn More

1-Tritylimidazole, 98%, Thermo Scientific Chemicals

Product Code. 11378636
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378636 5 g 5g
11368636 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378636 Supplier Thermo Scientific Alfa Aesar Supplier No. L17694.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is a synthesis material intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a synthesis material intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15469-97-3
Molecular Formula C22H18N2
Molecular Weight (g/mol) 310.40
MDL Number MFCD00229427
InChI Key NPZDCTUDQYGYQD-UHFFFAOYSA-N
Synonym 1-trityl-1h-imidazole, 1-triphenylmethyl imidazole, n-tritylimidazole, 1-triphenylmethyl-1h-imidazole, clotrimazole impurity f, triphenylmethyl imidazole, 1-triphenylmethylimidazole, 1-tritylimidazol, pubchem23733, acmc-209dbb
PubChem CID 618231
SMILES C1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 220°C to 224°C
Odor Odorless
Quantity 5 g
Beilstein 238171
Solubility Information Insoluble in water.
IUPAC Name 1-(triphenylmethyl)-1H-imidazole
Formula Weight 310.4
Percent Purity 98%
Chemical Name or Material 1-Tritylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.