missing translation for 'onlineSavingsMsg'
Learn More

10 μS/cm Conductivity Standard at 25 °C, Reagecon
BIOPHARMA

Product Code. 16460594 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460594 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16460594 Supplier Reagecon Supplier No. CSKC1025
  • ONLINE EXCLUSIVE PRICE
    143.00 €
    119.30 € / 250mL
    Save 23.70 €
    16% Off
View all promotions
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aqueous based, non-hazardous standard

  • Tested by ISO 17025 accredited test method
  • NIST traceable
  • Low temperature coefficient of variation
  • Guaranteed stability throughout entire shelf life even after opening bottle
  • Accuracy: 10 μS/cm ± 1%

Specifications

Packaging HDPE bottle
Traceability to NIST Yes
For Use With (Equipment) Conductivity Meter
Type Conductivity Standard
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.