missing translation for 'onlineSavingsMsg'
Learn More

100% Biodiesel, VHG™

Product Code. 17988564
Change view
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988564 100 mL 100mL
17937144 20 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17988564 Supplier VHG Labs Supplier No. VHGBDBLEND100P100
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

100% Biodiesel
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bio Diesel Fuel
CAS 870530-78-2
Analyte or Component Names Bio Diesel Fuel
Shipping Condition Room Temperature
Concentration 100%
Synonym Diesel fuel substitutes, biodiesel, B 10, B 10 biofuel, B 100, B 100 biofuel, B 20, B 20 biofuel, B 5, B 5 biofuel, B 50, B 50 biofuel, Biodiesel, Biofuels, biodiesel, Biodiesel fuel
IUPAC Name Biodiesel Fuel
Solvent or Matrix High Cetane Diesel Fuel/Biodiesel
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.