missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V21+K Wear Metals Standard: 900 μg/g Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG™
Click to view available options
Quantity:
50 g
100 g
200 g
400 g
Unit Size:
100g
200g
400g
50g
Description
V21+K Wear Metals Standard: 900 μg/g Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Aluminium/Barium/Boron/Cadmium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-39-3, 7440-42-8, 7440-43-9, 7440-70-2, 7440-47-3 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Aluminium/Barium/Boron/Cadmium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hydrocarbon Oil |
| Quantity | 100 g |
| Concentration or Composition (by Analyte or Components) | Aluminium 900 μg/g/Barium 900 μg/g/Boron 900 μg/g/Cadmium 900 μg/g/Calcium 900 μg/g/Chromium 900 μg/g/Copper 900 μg/g/Iron 900 μg/g/Lead 900 μg/g/Magnesium 900 μg/g/Manganese 900 μg/g/Molybdenum 900 μg/g/Nickel 900 μg/g/Phosphorus 900 μg/g/Potassium 900 μg/g/Silicon 900 μg/g/Silver 900 μg/g/Sodium 900 μg/g/Tin 900 μg/g/Titanium 900 μg/g/Vanadium 900 μg/g/Zinc 900 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction