missing translation for 'onlineSavingsMsg'
Learn More

C29, MedChemExpress

Product Code. 16434336
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434336

Brand: Medchem Express HY100461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress C29 is a Toll-like receptor 2 (TLR2) inhibitor. C29 blocks hTLR2/1 and hTLR2/6 signaling with IC50s of 19.7 and 37.6 μM, respectively.

C29 is a Toll-like receptor 2 (TLR2) inhibitor. C29 blocks hTLR2/1 and hTLR2/6 signaling with IC50s of 19.7 and 37.6 μM, respectively.

Specifications

Chemical Name or Material C29
CAS 363600-92-4
Color Orange
Purity Grade Notes Research
Molecular Formula C16H15NO4
Quantity 100 mg
Solubility Information DMSO : ≥ 30 mg/mL (105.16 mM)
SMILES OC1=C(/C=N/C2=C(C)C(C(O)=O)=CC=C2)C=CC=C1OC
Molecular Weight (g/mol) 285.29
Formula Weight 285.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.