missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspofungin diacetate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
25mg
500mg
50mg
5mg
Description
Caspofungin (MK-0991) diacetate is a potent antifungal agent. Caspofungin diacetate inhibits the synthesis of the fungal cell wall component β-(l,3)-D-glucan.
Specifications
Specifications
| Chemical Name or Material | Caspofungin diacetate |
| Physical Form | Solid |
| CAS | 179463-17-3 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C56H96N10O19 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | MK-0991 diacetateL-743872 diacetate |
| Solubility Information | H2O : ≥ 100 mg/mL (82.41 mM) ∣DMSO : 100 mg/mL (82.41 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction