missing translation for 'onlineSavingsMsg'
Learn More

Doramapimod, MedChemExpress

Product Code. 16464856
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16464856

Brand: Medchem Express HY10320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doramapimod (BIRB 796) is an orally active, highly potent p38 MAPK inhibitor, which has an IC50 for p38α=38 nM, for p38β=65 nM, for p38γ=200 nM, and for p38δ=520 nM. Doramapimod has picomolar affinity for p38 kinase (Kd=0.1 nM). Doramapimod also inhibits B-Raf with an IC50 of 83 nM.

Doramapimod (BIRB 796) is an orally active, highly potent p38 MAPK inhibitor, which has an IC50 for p38α=38 nM, for p38β=65 nM, for p38γ=200 nM, and for p38δ=520 nM. Doramapimod has picomolar affinity for p38 kinase (Kd=0.1 nM). Doramapimod also inhibits B-Raf with an IC50 of 83 nM.

Specifications

Chemical Name or Material Doramapimod
CAS 285983-48-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H37N5O3
Quantity 100 mg
Synonym BIRB 796
Solubility Information DMSO : 125 mg/mL (236.89 mM; Need ultrasonic) ∣Ethanol : 33.33 mg/mL (63.17 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C(C)(C)C)=NN1C2=CC=C(C)C=C2)NC3=C4C=CC=CC4=C(OCCN5CCOCC5)C=C3
Molecular Weight (g/mol) 527.66
Formula Weight 527.66
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.