missing translation for 'onlineSavingsMsg'
Learn More

DUPA, MedChemExpress

Product Code. 16475366
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475366 100 mg 100mg
16487217 5 mg 5mg
16470807 50 mg 50mg
16464498 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475366 Supplier Medchem Express Supplier No. HY111606

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DUPA, belongs to a class of glutamate ureas, is used as the targeting moiety in drug conjugate to selectively deliver cytotoxic drugs to prostate cancer cells.

DUPA, belongs to a class of glutamate ureas, is used as the targeting moiety in drug conjugate to selectively deliver cytotoxic drugs to prostate cancer cells.

Specifications

Chemical Name or Material DUPA
Physical Form Solid
CAS 302941-52-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H16N2O9
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 300 mg/mL (936.77 mM) ∣H2O : ≥ 150 mg/mL (468.38 mM)
SMILES O=C(N[C@](CCC(O)=O)([H])C(O)=O)N[C@](CCC(O)=O)([H])C(O)=O
Molecular Weight (g/mol) 320.25
Formula Weight 320.25
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.