missing translation for 'onlineSavingsMsg'
Learn More

Erlotinib Hydrochloride, MedChemExpress

Product Code. 16465686
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16465686

Brand: Medchem Express HY12008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Erlotinib Hydrochloride (CP-358774 Hydrochloride) inhibits purified EGFR kinase with an IC50 of 2 nM.

Erlotinib Hydrochloride (CP-358774 Hydrochloride) inhibits purified EGFR kinase with an IC50 of 2 nM.

Specifications

Chemical Name or Material Erlotinib Hydrochloride
CAS 183319-69-9
Color White
Purity Grade Notes Research
Molecular Formula C22H24ClN3O4
Quantity 100 mg
Synonym CP-358774 hydrochlorideNSC 718781 hydrochlorideOSI-774 hydrochloride
Solubility Information DMSO : 6.2 mg/mL (14.42 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES COCCOC1=CC2=NC=NC(NC3=CC=CC(C#C)=C3)=C2C=C1OCCOC.[H]Cl
Molecular Weight (g/mol) 429.9
Formula Weight 429.9
Percent Purity 99.71%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.