missing translation for 'onlineSavingsMsg'
Learn More

Glycopyrrolate, MedChemExpress

Product Code. 16427326
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427326 100 mg 100mg
16447358 10 mg 10mg
16473377 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. Se returpolicy
Artikelnummer. 16427326 Leverandør Medchem Express Leverandørnr. HY17465

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Glycopyrrolate (Glycopyrronium bromide) is a muscarinic competitive antagonist used as an antispasmodic.

Glycopyrrolate (Glycopyrronium bromide) is a muscarinic competitive antagonist used as an antispasmodic.

Tekniske data

Chemical Name or Material Glycopyrrolate
Physical Form Solid
CAS 596-51-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H28BrNO3
For Use With (Application) Neuroscience-Neuromodulation
Synonym Glycopyrronium bromideGlycopyrrolate bromide
Solubility Information H2O : 100 mg/mL (251.05 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (251.05 mM; Need ultrasonic)
SMILES C[N+]1(C)CC(OC(C(O)(C2CCCC2)C3=CC=CC=C3)=O)CC1.[Br-]
Molecular Weight (g/mol) 398.33
Formula Weight 398.33
Percent Purity 99.8%
Grade Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.