missing translation for 'onlineSavingsMsg'
Learn More

Nitazoxanide, MedChemExpress

Product Code. 16468176
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16468176

Brand: Medchem Express HYB0217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nitazoxanide (NTZ), an anthelmintic agent, exhibits a broad spectrum of activities against a wide variety of helminths, protozoa, and enteric bacteria infecting animals and humans.

Nitazoxanide inhibits Giardia lamblia trophozoite proliferation in axenic culture with an IC50 of 2.4 μM. Nitazoxanide can be used for the research of parasitic gastroenteritis. Nitazoxanide shows anti-Japanese encephalitis virus (JEV) activity in a mouse model.

Specifications

Chemical Name or Material Nitazoxanide
Physical Form Powder
CAS 55981-09-4
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H9N3O5S
For Use With (Application) COVID-19-anti-virus
Synonym NTZ NSC 697855
Solubility Information DMSO : ≥ 100 mg/mL (325.44 mM)
SMILES O=C(NC1=NC=C([N+]([O-])=O)S1)C2=CC=CC=C2OC(C)=O
Molecular Weight (g/mol) 307.28
Formula Weight 307.28
Percent Purity 98.35%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.